Learn More
Apexbio Technology LLC PSB 1114 500ug
SDP
Catalog No. 5000087883
Encompass_Preferred
PSB 1114 is a small-molecule agonist targeting the purinergic P2Y2 receptor It is designed to selectively activate the P2Y2 receptor a G protein-coupled receptor (GPCR) naturally activated by extracellular nucleotides thereby regulating downstream signaling pathways such as chloride ion secretion mucosal hydration and inflammatory responses PSB 1114 exerts its biological activity primarily through P2Y2 receptor activation It demonstrates subtype selectivity within the P2Y receptor family with an EC50 value of approximately 0 134 M for the P2Y2 receptor and exhibits lower affinity towards related subtypes Based on these pharmacological properties PSB 1114 holds research potential in studies exploring therapeutic approaches for respiratory diseases including cystic fibrosis via modulation of ion transport and mucosal hydration mechanisms mediated by P2Y2 receptor activation
Catalog No. 50-000-87883 Supplier Apexbio Technology LLC Supplier No. B76490.5
PSB 1114 is a small-molecule agonist targeting the purinergic P2Y2 receptor It is designed to selectively activate the P2Y2 receptor a G protein-coupled receptor (GPCR) naturally activated by extracellular nucleotides thereby regulating downstream signaling pathways such as chloride ion secretion mucosal hydration and inflammatory responses PSB 1114 exerts its biological activity primarily through P2Y2 receptor activation It demonstrates subtype selectivity within the P2Y receptor family with an EC50 value of approximately 0 134 M for the P2Y2 receptor and exhibits lower affinity towards related subtypes Based on these pharmacological properties PSB 1114 holds research potential in studies exploring therapeutic approaches for respiratory diseases including cystic fibrosis via modulation of ion transport and mucosal hydration mechanisms mediated by P2Y2 receptor activation