Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Psb-1114 triethylamine | 00-00-0 | 95.0% | C10H15F2N3O13P3S.xC6H15N | 5 MG
SDP

Catalog No. 5000263756
Encompass_Preferred

PSB-1114 triethylamine is a potent, enzymatically stable, subtype-selective agonist of the P2Y2 receptor with an EC50 of 134 nM. It demonstrates over 50-fold selectivity versus P2Y4 and P2Y6 and is provided at research-grade purity for laboratory studies.

  • Potent P2Y2 receptor agonist with an EC50 of 134 nM.
  • Enzymatically stable for biological assays.
  • Subtype-selective with >50-fold selectivity versus P2Y4 and P2Y6.
  • Supplied at 95.0% research purity.
  • Available in small research pack sizes such as 1 mg, 5 mg, 10 mg, and 50 mg.

Catalog No. 50-002-63756 Supplier Medchemexpress LLC Supplier No. HY110092A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSB-1114 triethylamine is a potent, enzymatically stable, subtype-selective agonist of the P2Y2 receptor with an EC50 of 134 nM. It demonstrates over 50-fold selectivity versus P2Y4 and P2Y6 and is provided at research-grade purity for laboratory studies.

  • Potent P2Y2 receptor agonist with an EC50 of 134 nM.
  • Enzymatically stable for biological assays.
  • Subtype-selective with >50-fold selectivity versus P2Y4 and P2Y6.
  • Supplied at 95.0% research purity.
  • Available in small research pack sizes such as 1 mg, 5 mg, 10 mg, and 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.