Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PSB-1114 triethylamine | 95.0% | 10 MG
SDP

Catalog No. 5000344944
Encompass_Preferred

PSB-1114 triethylamine is a potent, enzymatically stable, and subtype-selective P2Y2 receptor agonist with an EC50 of 134 nM. It displays >50-fold selectivity versus the P2Y4 (EC50 of 9.3 μM) and P2Y6 (EC50 of 7.0 μM) receptors.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: -20°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture).
  • Solubility (in vitro): H2O: ≥ 100 mg/mL

Catalog No. 50-003-44944 Supplier Medchemexpress LLC Supplier No. HY110092A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSB-1114 triethylamine is a potent, enzymatically stable, and subtype-selective P2Y2 receptor agonist with an EC50 of 134 nM. It displays >50-fold selectivity versus the P2Y4 (EC50 of 9.3 μM) and P2Y6 (EC50 of 7.0 μM) receptors.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: -20°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture).
  • Solubility (in vitro): H2O: ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.