Learn More
Apexbio Technology LLC PSB 1115 152529-79-8 10mg
SDP
Catalog No. 5000088571
Encompass_Preferred
PSB 1115 is a selective antagonist targeting the human A2B adenosine receptor It is designed to inhibit receptor-mediated signaling pathways associated with adenosine receptors thereby regulating processes related to inflammation and immune response PSB 1115 exerts its biological activity primarily through antagonism of the A2B adenosine receptor In experimental studies PSB 1115 demonstrates inhibition of acetylcholine-induced contractions triggered by 2 4 6-trinitrobenzene sulfonic acid (TNBS) a commonly used method in gastrointestinal inflammation research Based on these pharmacological properties PSB 1115 holds research potential in investigating inflammatory response immune regulation and mechanisms underlying inflammation-related disorders
Catalog No. 50-000-88571 Supplier Apexbio Technology LLC Supplier No. B692210
PSB 1115 is a selective antagonist targeting the human A2B adenosine receptor It is designed to inhibit receptor-mediated signaling pathways associated with adenosine receptors thereby regulating processes related to inflammation and immune response PSB 1115 exerts its biological activity primarily through antagonism of the A2B adenosine receptor In experimental studies PSB 1115 demonstrates inhibition of acetylcholine-induced contractions triggered by 2 4 6-trinitrobenzene sulfonic acid (TNBS) a commonly used method in gastrointestinal inflammation research Based on these pharmacological properties PSB 1115 holds research potential in investigating inflammatory response immune regulation and mechanisms underlying inflammation-related disorders