Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PSB-1115 | 152529-79-8 | 99.0% | 350.35 | C14H14N4O5S | 5 MG
SDP

Catalog No. 5000247457
Encompass_Preferred

PSB-1115 is a selective A2B adenosine receptor antagonist supplied for research use to investigate A2B-mediated signaling and physiological responses. It has documented activity in TNBS-induced contraction models and is provided in solid and solution formats for experimental applications.

  • Selective A2B adenosine receptor antagonist.
  • Research-use compound for A2B receptor studies.
  • Purity 99.0%.
  • Molecular formula C14H14N4O5S; molecular weight 350.35.
  • CAS number 152529-79-8.
  • Available in solid 5 mg and solution 10 mM 1 mL in DMSO.
  • Store at -20°C; protect from light. In solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-47457 Supplier Medchemexpress LLC Supplier No. HY1031825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSB-1115 is a selective A2B adenosine receptor antagonist supplied for research use to investigate A2B-mediated signaling and physiological responses. It has documented activity in TNBS-induced contraction models and is provided in solid and solution formats for experimental applications.

  • Selective A2B adenosine receptor antagonist.
  • Research-use compound for A2B receptor studies.
  • Purity 99.0%.
  • Molecular formula C14H14N4O5S; molecular weight 350.35.
  • CAS number 152529-79-8.
  • Available in solid 5 mg and solution 10 mM 1 mL in DMSO.
  • Store at -20°C; protect from light. In solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.