Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PSB-1115 | 152529-79-8 | 99.0% | 350.35 g·mol⁻¹ | C14H14N4O5S | 100 MG
SDP

Catalog No. 5000246464
Encompass_Preferred

PSB-1115 is a selective A2B adenosine receptor antagonist used as a research tool to block A2B-mediated signaling in vitro and in vivo. It is reported to antagonize adenosine-mediated effects in animal models and to inhibit TNBS-induced responses.

  • Selective A2B adenosine receptor antagonist for research use.
  • High purity (99.0%) solid suitable for biological assays.
  • Chemical formula C14H14N4O5S; molecular weight 350.35 g·mol⁻¹.
  • Soluble in DMSO (100 mg/mL) and water (10 mg/mL) with sonication or warming.
  • Recommended storage at -20°C; in solution, store at -80°C for long-term stability.
  • Reported in vivo activity at 0.5 mg/kg (i.p.) in mouse mucociliary clearance studies.

Catalog No. 50-002-46464 Supplier Medchemexpress LLC Supplier No. HY103182100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSB-1115 is a selective A2B adenosine receptor antagonist used as a research tool to block A2B-mediated signaling in vitro and in vivo. It is reported to antagonize adenosine-mediated effects in animal models and to inhibit TNBS-induced responses.

  • Selective A2B adenosine receptor antagonist for research use.
  • High purity (99.0%) solid suitable for biological assays.
  • Chemical formula C14H14N4O5S; molecular weight 350.35 g·mol⁻¹.
  • Soluble in DMSO (100 mg/mL) and water (10 mg/mL) with sonication or warming.
  • Recommended storage at -20°C; in solution, store at -80°C for long-term stability.
  • Reported in vivo activity at 0.5 mg/kg (i.p.) in mouse mucociliary clearance studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.