Learn More
TARGETMOL CHEMICALS INC PSB 1115 5MG
SDP
Catalog No. 502580891
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. PSB 1115 is an A2B receptor antagonist and can counteract the inhibitory effect of NECA. purity: 99%
Catalog No. 50-258-0891 Supplier TARGETMOL CHEMICALS INC Supplier No. T232015MG
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. PSB 1115 is an A2B receptor antagonist and can counteract the inhibitory effect of NECA. purity: 99%