Learn More
Medchemexpress LLC HY-101910 5mg Medchemexpress, PSB-12062 CAS:55476-47-6 Purity:>98%
SDP
Catalog No. 501967913
Encompass_Preferred
Medchemexpress, HY-101910 5mg PSB-12062 CAS:55476-47-6 PSB-12062 is a potent and selective P2X4 antagonist with an IC50 of 1.38 μM for human P2X4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7913 Supplier Medchemexpress LLC Supplier No. HY1019105MG
Medchemexpress, HY-101910 5mg PSB-12062 CAS:55476-47-6 PSB-12062 is a potent and selective P2X4 antagonist with an IC50 of 1.38 μM for human P2X4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.