Learn More
TARGETMOL CHEMICALS INC PSB-12379 DITRIETHYLAMIN 5MG
SDP
Catalog No. 502581439
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PSB-12379 ditriethylamine salt a nucleotide analogue is a potent Ecto-5'-Nucleotidase (CD73) inhibitor with Kis of 9.03 nM (rat) and 2.21 nM (human).
Catalog No. 50-258-1439 Supplier TARGETMOL CHEMICALS INC Supplier No. T12569L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PSB-12379 ditriethylamine salt a nucleotide analogue is a potent Ecto-5'-Nucleotidase (CD73) inhibitor with Kis of 9.03 nM (rat) and 2.21 nM (human).