Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PSB-603 | 1092351-10-4 | 99.4% | 529.01 g/mol | C24H25ClN6O4S | 25 MG
SDP

Catalog No. 5000247592
Encompass_Preferred

PSB-603 (CAS 1092351-10-4) is a potent, highly selective adenosine A2B receptor antagonist used in pharmacological research to block A2B-mediated signaling and investigate inflammatory responses. It exhibits subnanomolar affinity for A2B (Ki = 0.553 nM) and shows minimal activity at other adenosine receptor subtypes.

  • Potent A2B antagonist with Ki = 0.553 nM.
  • High purity (99.4%) as supplied.
  • Molecular formula C24H25ClN6O4S; molecular weight 529.01 g/mol.
  • Solid powder suitable for in vitro and in vivo studies.
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term); solutions at -80°C.

Catalog No. 50-002-47592 Supplier Medchemexpress LLC Supplier No. HY10316625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSB-603 (CAS 1092351-10-4) is a potent, highly selective adenosine A2B receptor antagonist used in pharmacological research to block A2B-mediated signaling and investigate inflammatory responses. It exhibits subnanomolar affinity for A2B (Ki = 0.553 nM) and shows minimal activity at other adenosine receptor subtypes.

  • Potent A2B antagonist with Ki = 0.553 nM.
  • High purity (99.4%) as supplied.
  • Molecular formula C24H25ClN6O4S; molecular weight 529.01 g/mol.
  • Solid powder suitable for in vitro and in vivo studies.
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term); solutions at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.