Learn More
Medchemexpress LLC PSB-KK1415 | 885896-26-4 | 99.9% | 462.93 | 25 MG
SDP
Catalog No. 5000395122
Encompass_Preferred
PSB-KK1415 is a selective agonist for human orphan G protein-coupled receptor GPR18, with an EC50 of 19.1 nM. This product is for research use only and not sold to patients.
  • Selective agonist for human orphan G protein-coupled receptor GPR18
  • Potentially a promising drug target for cancer and immunity
Catalog No. 50-003-95122 Supplier Medchemexpress LLC Supplier No. 25MGHY162617
PSB-KK1415 is a selective agonist for human orphan G protein-coupled receptor GPR18, with an EC50 of 19.1 nM. This product is for research use only and not sold to patients.
  • Selective agonist for human orphan G protein-coupled receptor GPR18
  • Potentially a promising drug target for cancer and immunity