Learn More
Medchemexpress LLC Psb-sb-487 | 1399049-81-0 | 98.0% | 408.53 g/mol | C26H32O4 | 50MG

Supplier: Medchemexpress LLC HY12511150MG
PSB-SB-487 is a small-molecule research compound that functions as a potent GPR55 antagonist and a CB2 receptor agonist. It is supplied as a white to off-white solid for laboratory use and is intended for research use only. The compound has CAS number 1399049-81-0 and a molecular weight of 408.53 g/mol.
- Potent GPR55 antagonist and CB2 agonist.
- High purity (approximately 98.0%).
- Molecular formula C26H32O4; molecular weight 408.53 g/mol.
- Supplied as a white to off-white solid.
- Soluble in DMSO at high concentration (100 mg/mL; ultrasonic recommended).
- Recommended storage conditions for powder and solutions to maintain stability.
- For research use only; not for human or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.