Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ PSC Cardiomyocyte Maintenance Medium
GREENER_CHOICE

Catalog No. A2920801
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2920801 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2920801 Supplier Gibco™ Supplier No. A2920801
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

PSC Cardiomyocyte Maintenance Medium

Gibco™ Cardiomyocyte Maintenance Medium is a serum-free and xeno-free medium that is capable of maintaining cardiomyocytes that have been differentiated using the PSC Cardiomyocyte Differentiation Kit (Cat. No. A2921201). This medium is included in the kit and sold separately for researchers wishing to maintain differentiated cardiomyocytes in culture for extended periods of time.
  • No thawing or mixing required—simply warm and add to differentiated cardiomyocyte culture
  • Long-term culture—maintain differentiated cardiomyocytes in culture for >15 days
  • Stable phenotype—maintain phenotype throughout long-term culture experiments
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Cardiomyocytes
Product Type Maintenance Medium
Serum Level Serum-free
Quantity 500 mL
Shipping Condition Room Temperature
Culture Type Mammalian Cell Culture
What substrate should I use for cardiomyocyte differentiation?

We recommend using Gibco Geltrex Flex LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix (Cat. No. A4000046801, A4000046802, or A4000046803) or Gibco Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A400457 or A400458) for xeno-free applications.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.