Learn More
Aviva Systems Biology Corp PSCA Antibody - C-terminal region (ARP42250_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42250_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSCA. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PSCA. Alternate names: PRO232ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-284-986