Learn More
ACROBiosystems ACROBiosystems Human PSCA Protein, His Tag
Catalog No. 501957003
Encompass_Preferred
Recombinant Protein;100UG;Human PSCA, His Tag (PSA-H52H6) is expressed from human 293 cells (HEK293). It contains AA Leu 12 - Ser 86 (Accession # O43653-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 10.4 kDa. The protein migrates as 18-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7003 Supplier ACROBiosystems Supplier No. PSAH52H6100UG
Recombinant Protein;100UG;Human PSCA, His Tag (PSA-H52H6) is expressed from human 293 cells (HEK293). It contains AA Leu 12 - Ser 86 (Accession # O43653-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 10.4 kDa. The protein migrates as 18-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.