Learn More
Enzo Life Sciences PSD 95 monoclonal antibody (6G6-1C9) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMPS002E
Encompass_Preferred
Clone: 6G6-1C9. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant rat PSD-95. UniProt: Q62108. Source: Purified from ascites. Species Reactivity: Mouse, Rat, Bovine. Applications: ICC, WB. Recommended dilutions: Immunocytochemistry (12.5µg/ml). Western Blot (1:250, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-667-830