Learn More
Meso Scale Discovery PSD-95 Kit
SDP
Catalog No. 5000079234
Encompass
The PSD-95 Kit from Mesoscale Discovery is a kit for studying the postsynaptic density protein PSD-95 The kit contains a PSD-95 protein standard a PSD-95 capture antibody a PSD-95 detection antibody and reagents for performing a sandwich ELISA to quantify PSD-95 levels in cell or tissue samples The capture and detection antibodies allow sensitive and specific detection of PSD-95 The kit can be used to study synaptic plasticity by measuring changes in PSD-95 levels
Catalog No. 50-000-79234 Supplier Meso Scale Discovery Supplier No. K150QND4
Add to Cart
Add to Cart
Request a Quote
The PSD-95 Kit from Mesoscale Discovery is a kit for studying the postsynaptic density protein PSD-95 The kit contains a PSD-95 protein standard a PSD-95 capture antibody a PSD-95 detection antibody and reagents for performing a sandwich ELISA to quantify PSD-95 levels in cell or tissue samples The capture and detection antibodies allow sensitive and specific detection of PSD-95 The kit can be used to study synaptic plasticity by measuring changes in PSD-95 levels