Learn More
Meso Scale Discovery PSD-95 Kit
SDP
Catalog No. 5000079233
Encompass
The PSD-95 Kit from Meso Scale Discovery is a 96-well plate assay kit for the quantitative measurement of PSD-95 protein in human and rodent samples The kit contains a 96-well plate coated with a PSD-95 capture antibody a detection antibody and buffers and reagents needed to run the assay The electrochemiluminescent detection allows for high sensitivity and a broad dynamic range The total assay time is under 3 hours This assay can be used to measure PSD-95 levels in cell and tissue lysates and biological fluids It provides a tool to study synaptic plasticity and neurological diseases
Catalog No. 50-000-79233 Supplier Meso Scale Discovery Supplier No. K150QND2
The PSD-95 Kit from Meso Scale Discovery is a 96-well plate assay kit for the quantitative measurement of PSD-95 protein in human and rodent samples The kit contains a 96-well plate coated with a PSD-95 capture antibody a detection antibody and buffers and reagents needed to run the assay The electrochemiluminescent detection allows for high sensitivity and a broad dynamic range The total assay time is under 3 hours This assay can be used to measure PSD-95 levels in cell and tissue lysates and biological fluids It provides a tool to study synaptic plasticity and neurological diseases