Learn More
AdipoGen anti-PSD-95 (palmitoylated), mAb (rec.) (PF11) (100 µg)
SDP
Supplier:  AdipoGen AG27B0021C100
Encompass_Preferred
Recombinant Antibody. Recognizes human, mouse and rat palmitoylated PSD-95. Isotype: Human IgG2. Clone: PF11. Applications: ICC, IHC. Liquid. In PBS containing 10% glycerol. PSD-95 is a member of proteins located at a specialized postsynaptic membrane region, called the postsynaptic density (PSD). PSD-95 is the most abundant scaffold protein specifically enriched in the PSD. Through its PDZ domains, PSD-95 assembles various synaptic components at the PSD including intracellular signaling molecules (e.g. SynGAP and kalirin-7), ion channels and cell adhesion molecules (e.g. neuroligin). PSD-95 plays a primary role in synaptic development and maturation and is regulated by palmitoylation at its N-terminal cysteine residues leading to its postsynaptic targeting. Palmitoylation of PSD-95 is triggered by DHHC2, a plasma membrane-inserted palmitoylating enzyme. Palmitoylated PSD-95 is almost exclusively localized at excitatory synapses in neurons.
Catalog No. 50-205-3045