Learn More
Aviva Systems Biology Corp PSD3 Antibody - middle region (ARP42443_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42443_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PSD3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human PSD3. Alternate names: DKFZp761K1423, EFA6R, HCA67ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53334