Learn More
Enzo Life Sciences PSD95 monoclonal antibody (7E3-1B8) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMPS001E
Encompass_Preferred
Clone: 7E3-1B8. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant rat PSD-95. UniProt ID: Q62108. Source: Purified from ascites. Species reactivity: Mouse, Rat, Bovine. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7685