Learn More
Cayman Chemical PSelectIn GlycoproteIn LIgn
SDP
Supplier:  Cayman Chemical 38062100 UG
Encompass_Preferred
Source: Recombinant C-terminal human IgG1 Fc-His-tagged PSGL-1 extracellular domain expressed in HEK293 cells • Amino acids: 18-295 • MW: 57.1 kDa
Catalog No. 50-291-7723