Learn More
Cayman Chemical PSelectIn GlycoproteIn LIgn
SDP
Catalog No. 502915812
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged PSGL-1 expressed in HEK293 cells • Amino acids: 18-295 • MW: 30.5 kDa
Catalog No. 50-291-5812 Supplier Cayman Chemical Supplier No. 38072100 UG
Source: Recombinant human C-terminal His-tagged PSGL-1 expressed in HEK293 cells • Amino acids: 18-295 • MW: 30.5 kDa