Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human P-Selectin Protein, Fc Tag, 1 mg

Catalog No. 501658925
Encompass_Preferred

Human P-Selectin (CD62P) is a cell adhesion molecule expressed on activated endothelial cells and platelets. It functions by binding to ligands such as P-selectin glycoprotein ligand-1 (PSGL-1) to mediate leukocyte recruitment and is involved in tumor metastasis. This protein is expressed in human 293 cells and contains a human IgG1 Fc tag at the C-terminus.

  • Purity: >95% as determined by SDS-PAGE
  • Molecular weight: 106.6 kDa (calculated); migrates as 130-180 kDa due to glycosylation
  • Endotoxin: <1.0 EU per μg
  • Formulation: Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5 with trehalose
  • Stability: 12 months at -20°C to -70°C in lyophilized state; 3 months at -70°C after reconstitution
  • Storage: Avoid repeated freeze-thaw cycles

Catalog No. 50-165-8925 Supplier ACROBiosystems Supplier No. SEPH52551MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human P-Selectin (CD62P) is a cell adhesion molecule expressed on activated endothelial cells and platelets. It functions by binding to ligands such as P-selectin glycoprotein ligand-1 (PSGL-1) to mediate leukocyte recruitment and is involved in tumor metastasis. This protein is expressed in human 293 cells and contains a human IgG1 Fc tag at the C-terminus.

  • Purity: >95% as determined by SDS-PAGE
  • Molecular weight: 106.6 kDa (calculated); migrates as 130-180 kDa due to glycosylation
  • Endotoxin: <1.0 EU per μg
  • Formulation: Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5 with trehalose
  • Stability: 12 months at -20°C to -70°C in lyophilized state; 3 months at -70°C after reconstitution
  • Storage: Avoid repeated freeze-thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.