Learn More
Medchemexpress LLC HY-112217A 5mg Medchemexpress, PSEM 89S (TFA) CAS:1336913-03-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112217A5MG
Encompass_Preferred
Medchemexpress, HY-112217A 5mg PSEM 89S (TFA) CAS:1336913-03-1 PSEM 89S TFA is a selective and brain penetrant agonists for the resulting ion channels. PSEM 89S TFA is orthogonally selective for Q79G and L141F, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0811