Learn More
Aviva Systems Biology Corp PSEN1 Antibody - middle region (ARP58941_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58941_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSEN1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PSEN1. Alternate names: AD3, FAD, PS1, S182, PS-1ApplicationIHC, IP, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-535