Learn More
Aviva Systems Biology Corp Psenen Antibody - N-terminal region (ARP59440_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59440_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Psenen. It was validated on Western Blot. . Alternate names: 1700023M09Rik, MGC102026ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-696