Learn More
Aviva Systems Biology Corp PSENEN Antibody - C-terminal region (ARP59439_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59439_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSENEN. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PSENEN. Alternate names: MDS033, MSTP064, PEN-2, PEN2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-695