Learn More
American Radiolabeled Chemicals Inc PSER L-DIOL-1-14C 10UC
SDP
Catalog No. 50352979
Encompass_Preferred
Phosphatidylserine, L-a-dioleoyl-oleoyl-1-14C, 10uCi in ethanol:toluene (1:1), 0.1 mCi/ml, 50-60 mCi/mmol, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-979 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 363510UC
Phosphatidylserine, L-a-dioleoyl-oleoyl-1-14C, 10uCi in ethanol:toluene (1:1), 0.1 mCi/ml, 50-60 mCi/mmol, Shipped in Dry Ice, Exclusive