Learn More
CPC Scientific H-Ala-Arg-Thr-Lys-Gln-Thr-Ala-Arg-Lys-Ser(PO3H2)-Thr-Gly-Gly-Lys-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST023A
Encompass
This peptide is the fragment of Histone H3 (1-15) phosphorylated at Ser10. ONE-LETTER SEQUENCE: ARTKQTARK-pS-TGGKAMOLECULAR FORMULA: MOLECULAR WEIGHT:1640.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9456