Learn More
Enzo Life Sciences [pSer185]14-3-3 polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA4790025
Encompass_Preferred
Immunogen: 14-3-3 phosphorylated at Ser185. Species reactivity: Human, Mouse, Rat, Sheep, Bovine. Specificity: Recognizes 14-3-3β and 14-3-3ζ phosphorylated at Ser185. Applications: WB. Purity: Affinity purified phospho-specific antibody.
Catalog No. 50-201-1835