Learn More
CPC Scientific H-Ala-Thr-Lys-Ala-Ala-Arg-Lys-Ser(PO3H2)-Ala-Pro-Ser-Thr-Gly-Gly-Val-Lys-Lys-Pro-His-Arg-Tyr-Arg-Pro-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST056B
Encompass
This peptide is the fragment of Histone H3 (21-44) phosphorylate at Ser28. Phosphorylation of Ser28 occurs in prophase of mitosis and is associated with the initiation of chromosome condensation.ONE-LETTER SEQUENCE: ATKAARK-pS-APSTGGVKKPHRYRPGMOLECULAR FORMULA: MOLECULAR WEIGHT:2601.9STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Goto, H. et al. Genes Cells 7, 11 (2002); Goto, H. et al. J Biol Chem 274, 25543 (1999)
Catalog No. 50-210-9504