Learn More
Enzo Life Sciences [pSer326]HSF1 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA902F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the N-terminus of human HSF1 phosphorylated at Ser 326. Source: Purified from rabbit serum. Species reactivity: Human. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7586