Learn More
CPC Scientific H-Lys-Asp-Thr-Lys-Pro-Thr-Pro-Thr-Lys-Gly-Lys-Ala-Ala-Ser(PO3H2)-Ala-Ser-Thr-Thr-Pro-Val-Lys-Lys-Asp-Val-Thr-Pro-Val-Lys-Ala-Asp-Thr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST009B
Encompass
Phosphorylation at histone H1 Ser42 occurs only after phosphorylation at Thr34, Thr46, or Thr53 in vivo. This peptide is histone H1 (29-59) phosphorylated at Ser42. Histone H1 phosphorylation is associated with chromosome condensation and transcriptional regulation.ONE-LETTER SEQUENCE: KDTKPTPTKGKAA-pS-ASTTPVKKDVTPVKADTMOLECULAR FORMULA: MOLECULAR WEIGHT:3249.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Garcia, B. et al. Mol Cell Proteomics 5, 1593 (2006) ; Dou, Y. and MA. Gorovsky Mol Cell 6, 225 (2000) ; Mizzen, C. et al. J Biol Chem 274, 14533 (1999); Swank, R. et al. Biochem 36, 13761 (1997)
Catalog No. 50-210-9439