Learn More
Hach Company Pseudomonas Broth, Plastic Ampules, pk/50
Supplier:  Hach Company 2812250
Encompass
Pseudomonas broth plastic ampules, 2 mL. For detection of Pseudomonas species. pk/50.
Catalog No. 50-199-6711