Learn More
AdipoGen Pseudoanguillosporin A
SDP
Supplier:  AdipoGen BVT0311M001
Encompass_Preferred
Chemical. CAS 1159392-22-9. Formula C17H26O3. MW 278.4. Isolated from Strobilurus sp. Antifungal. Respiration inhibitor. Inhibits mitochondrial respiration in fungi. Binds at the Q0-centre on cytochrome b and blocks the electron transfer between cytochrome b and cytochrome c1.
Catalog No. 50-256-1994