Learn More
Medchemexpress LLC Pseudoerythromycin A enol ether (LY267108) | 105882-69-7 | 99.5% | 715.91 | 1 MG
SDP
Catalog No. 5000439457
Encompass_Preferred
Pseudoerythromycin A enol ether is a degradation product of Erythromycin. It has no significant antimicrobial activity.
  • Increases lower esophageal sphincter (LES) pressure in cats.
Catalog No. 50-004-39457 Supplier Medchemexpress LLC Supplier No. HY1120571MG
Pseudoerythromycin A enol ether is a degradation product of Erythromycin. It has no significant antimicrobial activity.
  • Increases lower esophageal sphincter (LES) pressure in cats.