Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pseudoginsenoside RT5 | 98474-78-3 | MFCD10566641 | 99.3% | 654.87 g/mol | C36H62O10 | 10 MG
SDP

Catalog No. 5000296615
Encompass_Preferred

Pseudoginsenoside RT5 is a ginsenoside-derived natural product isolated from Panax quinquefolium, supplied as a high-purity analytical standard for research and analytical applications.

  • Purity 99.32%.
  • CAS number 98474-78-3.
  • Molecular weight 654.87 g/mol.
  • Chemical formula C36H62O10.
  • Pack size 10 MG (also available in 1 MG and 5 MG).
  • Intended for analytical and research use.

Catalog No. 50-002-96615 Supplier Medchemexpress LLC Supplier No. HYN054210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pseudoginsenoside RT5 is a ginsenoside-derived natural product isolated from Panax quinquefolium, supplied as a high-purity analytical standard for research and analytical applications.

  • Purity 99.32%.
  • CAS number 98474-78-3.
  • Molecular weight 654.87 g/mol.
  • Chemical formula C36H62O10.
  • Pack size 10 MG (also available in 1 MG and 5 MG).
  • Intended for analytical and research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.