Learn More
Enzo Life Sciences Pseudohypericin (1mg). CAS: 55954-61-5
SDP
Catalog No. 502008820
Encompass_Preferred
Inhibitor of protein kinase C (PKC) (IC50=15µg/ml). Antiviral and antiretroviral activity. Potent inhibitor of dopamine-β-hydroxylase (IC50=3µg/ml). Purity: ≥98% (HPLC, TLC). Appearance: Black to black-red powder. Solubility: Soluble in methanol, 100% ethanol, DMSO, acetone and other organic solvents. Long Term Storage: +4°C.
Catalog No. 50-200-8820 Supplier Enzo Life Sciences Supplier No. ALX350276M001
Inhibitor of protein kinase C (PKC) (IC50=15µg/ml). Antiviral and antiretroviral activity. Potent inhibitor of dopamine-β-hydroxylase (IC50=3µg/ml). Purity: ≥98% (HPLC, TLC). Appearance: Black to black-red powder. Solubility: Soluble in methanol, 100% ethanol, DMSO, acetone and other organic solvents. Long Term Storage: +4°C.