Learn More
Enzo Life Sciences Pseudohypericin (5mg). CAS: 55954-61-5
SDP
Supplier:  Enzo Life Sciences ALX350276M005
Encompass_Preferred
Inhibitor of protein kinase C (PKC) (IC50=15µg/ml). Antiviral and antiretroviral activity. Potent inhibitor of dopamine-β-hydroxylase (IC50=3µg/ml). Purity: ≥98% (HPLC, TLC). Solubility: Soluble in methanol, 100% ethanol, DMSO, acetone and other organic solvents. Long Term Storage: +4°C.
Catalog No. 50-200-8821