Learn More
Medchemexpress LLC Pseudolaric acid A | 82508-32-5 | MFCD31560788 | 99.9% | 388.45 g/mol | C22H28O6 | 10 MG
SDP
Catalog No. 5000295926
Encompass_Preferred
Pseudolaric Acid A is a diterpene acid isolated from the bark of Pseudolarix kaempferi and supplied as a high-purity research chemical for in vitro biological testing and analytical use. It has reported antifungal, cytotoxic, and antifertility activities.
  • High purity (99.9%) suitable for analytical and bioassay applications.
  • Available as solid and as a 10 mM solution in DMSO for convenient dosing.
  • Molecular formula C22H28O6 and molecular weight 388.45 g/mol for precise calculations.
  • Intended for research use only; not for human or clinical use.
  • Typically supplied with product documentation such as SDS and COA from the supplier.
Catalog No. 50-002-95926 Supplier Medchemexpress LLC Supplier No. HYN067310MG
Pseudolaric Acid A is a diterpene acid isolated from the bark of Pseudolarix kaempferi and supplied as a high-purity research chemical for in vitro biological testing and analytical use. It has reported antifungal, cytotoxic, and antifertility activities.
  • High purity (99.9%) suitable for analytical and bioassay applications.
  • Available as solid and as a 10 mM solution in DMSO for convenient dosing.
  • Molecular formula C22H28O6 and molecular weight 388.45 g/mol for precise calculations.
  • Intended for research use only; not for human or clinical use.
  • Typically supplied with product documentation such as SDS and COA from the supplier.