Learn More
AdipoGen Pseudolaric acid B (100 µg)
SDP
Supplier:  AdipoGen AGCN20083C100
Encompass_Preferred
Chemical. CAS: 82508-31-4. Formula: C23H28O8. MW: 432.5. Isolated from Pseudolarix kaempferi. Antifungal and antifertility compound. Antitumor compound. PPARalpha signaling agonist. Angiogenesis inhibitor. Apoptosis and autophagy inducer. Microtubule-destabilizing agent. Anti-inflammatory. Inhibits NF-kappaB and p38 signaling. Immunosuppressive.
Catalog No. 50-205-3438