Learn More
Cayman Chemical Pseudolaric Acid B, Purity: ≥98%, CAS: 82508-31-4, 10 mg
SDP
Catalog No. 502955358
Encompass_Preferred
A diterpene acid isolated from the bark of P. kaempferi, a traditional Chinese medicinal plant; has antifungal activities and diverse effects that are relevant to cancer therapy
Catalog No. 50-295-5358 Supplier Cayman Chemical Supplier No. 1352710 MG
A diterpene acid isolated from the bark of P. kaempferi, a traditional Chinese medicinal plant; has antifungal activities and diverse effects that are relevant to cancer therapy