Learn More
Enzo Life Sciences Pseudolaric acid B (1mg). CAS: 82508-31-4
SDP
Supplier:  Enzo Life Sciences ALX350108M001
Encompass_Preferred
Activates PPARα and the phospholipase C (PLC) signalling pathway. Stimulates peroxisomal fatty acyl-CoA oxidase activity. These effects can be blocked by staurosporine (ALX-380-014). Exhibits significant cytotoxic activities against numerous tumor cell lines. Binds tubulin. Inhibits angiogenesis. Reduces HIF-1α protein levels. Induces apoptosis. Antifungal. Alternative name: Pseudolarix acid B, PLAB, PAB. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO, 100% ethanol or chloroform. Long Term Storage: +4°C.
Catalog No. 50-200-8735