Learn More
Medchemexpress LLC HY-N6939 5mg Medchemexpress, Pseudolaric Acid B CAS:82508-31-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN69395MG
Encompass_Preferred
Medchemexpress, HY-N6939 5mg Pseudolaric Acid B CAS:82508-31-4 Pseudolaric Acid B is a diterpene isolated from the root of Pseudolarix kaempferi Gorden (pinaceae), has anti-cancer, antifungal, and antifertile activities, and shows immunosuppressive activity on T lymphocytes [1] [2] [3] . Pseudolaric Acid B inhibits hepatitis B virus (HBV) secretion through apoptosis and cell cycle arrest. Pseudolaric Acid B induces autophagy [4] [5] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3973