Learn More
Medchemexpress LLC Pseudolaric Acid B | 82508-31-4 | 5 MG
SDP
Catalog No. 5000418359
Encompass_Preferred
Pseudolaric Acid B is an orally active diterpene acid that exhibits anti-fungal, anti-fertility, anti-angiogenesis, and anticancer activities. It can induce tumor cell apoptosis and autophagy, inhibit hepatitis B virus (HBV) secretion, and has an immunosuppressive effect by selectively inhibiting T lymphocyte proliferation and IL-2 production.
  • Induces tumor cell apoptosis
  • Induces autophagy
  • Inhibits hepatitis B virus (HBV) secretion
  • Has immunosuppressive effect
  • Selectively inhibits the proliferation of T lymphocytes
  • Selectively inhibits the production of IL-2
  • Exhibits anti-fungal activity
  • Exhibits anti-fertility activity
  • Exhibits anti-angiogenesis activity
  • Exhibits anticancer activity
Catalog No. 50-004-18359 Supplier Medchemexpress LLC Supplier No. HYN69385MG
Pseudolaric Acid B is an orally active diterpene acid that exhibits anti-fungal, anti-fertility, anti-angiogenesis, and anticancer activities. It can induce tumor cell apoptosis and autophagy, inhibit hepatitis B virus (HBV) secretion, and has an immunosuppressive effect by selectively inhibiting T lymphocyte proliferation and IL-2 production.
  • Induces tumor cell apoptosis
  • Induces autophagy
  • Inhibits hepatitis B virus (HBV) secretion
  • Has immunosuppressive effect
  • Selectively inhibits the proliferation of T lymphocytes
  • Selectively inhibits the production of IL-2
  • Exhibits anti-fungal activity
  • Exhibits anti-fertility activity
  • Exhibits anti-angiogenesis activity
  • Exhibits anticancer activity