Learn More
Medchemexpress LLC Pseudolaric Acid B | 82508-31-4 | 50 MG
SDP
Catalog No. 5000417963
Encompass_Preferred
Pseudolaric Acid B is an orally active diterpene acid. It exhibits anti-fungal, anti-fertility, anti-angiogenesis, and anticancer activities, and can induce tumor cell apoptosis and autophagy. Additionally, Pseudolaric Acid B can inhibit the secretion of hepatitis B virus (HBV) and possesses an immunosuppressive effect, selectively inhibiting the proliferation of T lymphocytes and the production of IL-2.
  • Exhibits anti-fungal activity
  • Exhibits anti-fertility activity
  • Exhibits anti-angiogenesis activity
  • Exhibits anticancer activity
  • Induces tumor cell apoptosis
  • Induces autophagy in tumor cells
  • Inhibits hepatitis B virus (HBV) secretion
  • Possesses immunosuppressive effects
  • Selectively inhibits T lymphocyte proliferation
  • Selectively inhibits IL-2 production
  • Targets HIF-1α
Catalog No. 50-004-17963 Supplier Medchemexpress LLC Supplier No. HYN693950MG
Pseudolaric Acid B is an orally active diterpene acid. It exhibits anti-fungal, anti-fertility, anti-angiogenesis, and anticancer activities, and can induce tumor cell apoptosis and autophagy. Additionally, Pseudolaric Acid B can inhibit the secretion of hepatitis B virus (HBV) and possesses an immunosuppressive effect, selectively inhibiting the proliferation of T lymphocytes and the production of IL-2.
  • Exhibits anti-fungal activity
  • Exhibits anti-fertility activity
  • Exhibits anti-angiogenesis activity
  • Exhibits anticancer activity
  • Induces tumor cell apoptosis
  • Induces autophagy in tumor cells
  • Inhibits hepatitis B virus (HBV) secretion
  • Possesses immunosuppressive effects
  • Selectively inhibits T lymphocyte proliferation
  • Selectively inhibits IL-2 production
  • Targets HIF-1α