Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pseudolaric acid C | 82601-41-0 | MFCD16660657 | 99.7% | 390.43 | C21H26O7 | 10 MG
SDP

Catalog No. 5000295590
Encompass_Preferred

Pseudolaric acid C is a diterpenoid isolated from the root bark of Pseudolarix amabilis with reported antifungal activity. It is provided as a research-grade chemical for in vitro studies and analytical applications.

  • High purity reported at approximately 99.7%.
  • Molecular weight 390.43 g/mol.
  • CAS number 82601-41-0 for unambiguous identification.
  • Available in small research pack sizes, for example 10 MG or as a 10 mM solution in DMSO (1 ML).
  • Storage: powder -20°C (long term); in solvent -80°C (up to 6 months) or -20°C (short term).
  • Suitable for antifungal and biochemical research applications; not for human or veterinary use.

Catalog No. 50-002-95590 Supplier Medchemexpress LLC Supplier No. HYN067210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pseudolaric acid C is a diterpenoid isolated from the root bark of Pseudolarix amabilis with reported antifungal activity. It is provided as a research-grade chemical for in vitro studies and analytical applications.

  • High purity reported at approximately 99.7%.
  • Molecular weight 390.43 g/mol.
  • CAS number 82601-41-0 for unambiguous identification.
  • Available in small research pack sizes, for example 10 MG or as a 10 mM solution in DMSO (1 ML).
  • Storage: powder -20°C (long term); in solvent -80°C (up to 6 months) or -20°C (short term).
  • Suitable for antifungal and biochemical research applications; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.