Learn More
AdipoGen Pseudolaric acid B (1 mg)
SDP
Supplier:  AdipoGen CDXP0453M001
Encompass_Preferred
Chemical. CAS: 82508-31-4. Formula: C23H28O8. MW: 432.46. Pseudolaric acid B is a natural diterpenoid isolated from the bark of Pseudolarix kaempferi. It has anti-inflammatory, anticancer, immunosuppressive, antifungal and antifertility properties. Effects relevant to its anticancer properties include, apoptosis and autophagy induction, preventing angiogenesis, microtubule-destabilizing, induction of cell cycle arrest, inhibition of multidrug resistance and inhibition of tumor growth in vivo. It inhibits NF-kappa and p38 signaling and has been shown to be a PPARalpha signaling agonist.
Catalog No. 50-205-4610