Learn More
ATCC Pseudomonas aeruginosa
Catalog No. 502382213
Encompass
Pseudomonas aeruginosa strain PRD-10 [CIP 103467, NCIB 10421, PCI 812] is a whole genome-sequenced strain that was isolated from a water bottle in an animal room. This bacterium can be used in disinfectant testing, antimicrobial preservative assays, testing of antimicrobial handwashing formulations, testing of slimicide, and quality control of Biosynth products. It is recommended by ATCC for use in the test described in ASTM Standard Test Method E686-91 where only the taxon is specified.
Catalog No. 50-238-2213 Supplier ATCC Supplier No. 15442
Pseudomonas aeruginosa strain PRD-10 [CIP 103467, NCIB 10421, PCI 812] is a whole genome-sequenced strain that was isolated from a water bottle in an animal room. This bacterium can be used in disinfectant testing, antimicrobial preservative assays, testing of antimicrobial handwashing formulations, testing of slimicide, and quality control of Biosynth products. It is recommended by ATCC for use in the test described in ASTM Standard Test Method E686-91 where only the taxon is specified.